Assay Details
| Gene Symbol: CCDC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417442.1 | XP_022273150.1 | 1 - 2 | 627 | 115 | |
| XM_022417443.1 | XP_022273151.1 | 1 - 2 | 630 | 115 | ||
| XM_546111.6 | XP_546111.3 | 1 - 2 | 626 | 115 | ||
More Information
Set Membership: |
|

CCDC6
Genomic Map