Assay Details
| Gene Symbol: FOSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405633.1 | XP_022261341.1 | 5 - 6 | 1243 | 94 | |
| XM_022405634.1 | XP_022261342.1 | 3 - 4 | 1194 | 94 | ||
| XM_540845.6 | XP_540845.3 | 4 - 5 | 1155 | 94 | ||
More Information
Set Membership: |
|

FOSL1
Genomic Map