Assay Details
| Gene Symbol: CANT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624035.3 | XP_005624092.1 | 2326 | 164 | ||
| XM_005624037.2 | XP_005624094.1 | 1523 | 164 | |||
| XM_022422660.1 | XP_022278368.1 | 2488 | 164 | |||
| XM_540464.5 | XP_540464.2 | 1498 | 164 | |||
More Information
Set Membership: |
|

CANT1
Genomic Map