Assay Details
| Gene Symbol: EPM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417819.1 | XP_022273527.1 | 1061 | 137 | ||
| XM_022417820.1 | XP_022273528.1 | 727 | 137 | |||
| XM_022417821.1 | XP_022273529.1 | 1058 | 137 | |||
| XM_022417822.1 | XP_022273530.1 | 1057 | 137 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EPM2A
Genomic Map