Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014108587.2 | XP_013964062.1 | 4 | 634 | 108 | |
| XM_022410901.1 | XP_022266609.1 | 4 | 582 | 108 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

YAF2
Genomic Map