Assay Details
| Gene Symbol: MPHOSPH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419124.1 | XP_022274832.1 | 3 - 4 | 331 | 163 | |
| XM_536767.6 | XP_536767.2 | 3 - 4 | 349 | 163 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MPHOSPH6
Genomic Map