Assay Details
| Gene Symbol: POP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621028.3 | XP_005621085.1 | 447 | 91 | ||
| XM_005621029.3 | XP_005621086.1 | 490 | 91 | |||
| XM_005621030.3 | XP_005621087.1 | 426 | 91 | |||
| XM_022419635.1 | XP_022275343.1 | 443 | 91 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

POP7
Genomic Map