Assay Details
| Gene Symbol: CD2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621332.3 | XP_005621389.1 | 3 - 4 | 268 | 119 | |
| XM_022419860.1 | XP_022275568.1 | 2 - 3 | 266 | 119 | ||
| XM_022419861.1 | XP_022275569.1 | 2 - 3 | 258 | 119 | ||
| XM_547057.6 | XP_547057.2 | 2 - 3 | 258 | 119 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CD2BP2
Genomic Map