Assay Details
| Gene Symbol: DNAJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122027.1 | XP_013977502.1 | 3 - 4 | 810 | 145 | |
| XM_847807.5 | XP_852900.1 | 2 - 3 | 887 | 145 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DNAJB1
Genomic Map