Assay Details
| Gene Symbol: PAX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623336.3 | XP_005623393.1 | 4 | 1117 | 104 | |
| XM_022421937.1 | XP_022277645.1 | 4 | 1118 | 104 | ||
| XM_022421938.1 | XP_022277646.1 | 4 | 1116 | 104 | ||
| XM_022421939.1 | XP_022277647.1 | 4 | 1118 | 104 | ||
More Information
Set Membership: |
|

PAX9
Genomic Map