Assay Details
| Gene Symbol: RAD21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627881.3 | XP_005627938.1 | 187 | 110 | ||
| XM_022426666.1 | XP_022282374.1 | 187 | 110 | |||
| XM_539142.6 | XP_539142.2 | 368 | 110 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAD21
Genomic Map