Assay Details
| Gene Symbol: PCGF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014112695.2 | XP_013968170.1 | 10 - 11 | 969 | 88 | |
| XM_014112696.2 | XP_013968171.1 | 9 - 10 | 888 | 88 | ||
| XM_014112697.2 | XP_013968172.1 | 8 - 9 | 803 | 88 | ||
More Information
Set Membership: |
|

PCGF3
Genomic Map