Assay Details
| Gene Symbol: ZBTB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622598.3 | XP_005622655.1 | 3 | 1270 | 70 | |
| XM_022421144.1 | XP_022276852.1 | 2 | 1186 | 70 | ||
| XM_547497.6 | XP_547497.2 | 2 | 1997 | 70 | ||
More Information
Set Membership: |
|

ZBTB18
Genomic Map