Assay Details
| Gene Symbol: HTATIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633756.3 | XP_005633813.1 | 2 - 3 | 260 | 109 | |
| XM_534088.6 | XP_534088.3 | 2 - 3 | 329 | 109 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HTATIP2
Genomic Map