Assay Details
| Gene Symbol: CDC42EP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404642.1 | XP_022260350.1 | 2 | 646 | 146 | |
| XM_532935.6 | XP_532935.1 | 2 | 460 | 146 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDC42EP3
Genomic Map