Assay Details
| Gene Symbol: PAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113100.2 | XP_013968575.1 | 3 - 4 | 494 | 130 | |
| XM_022417851.1 | XP_022273559.1 | 3 - 4 | 495 | 130 | ||
| XM_022417852.1 | XP_022273560.1 | 3 - 4 | 506 | 130 | ||
| XM_022417853.1 | XP_022273561.1 | 3 - 4 | 486 | 130 | ||
| XM_022417854.1 | XP_022273562.1 | 3 - 4 | 523 | 130 | ||
| XM_022417855.1 | XP_022273563.1 | 3 - 4 | 490 | 130 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PAIP1
Genomic Map