Assay Details
| Gene Symbol: MXD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638977.4 | XP_003639025.1 | 4 - 5 | 404 | 86 | |
| XM_005618522.3 | XP_005618579.1 | 3 - 4 | 374 | 86 | ||
| XM_022417128.1 | XP_022272836.1 | 5 - 6 | 617 | 86 | ||
| XM_022417129.1 | XP_022272837.1 | 5 - 6 | 569 | 86 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MXD4
Genomic Map