Assay Details
| Gene Symbol: POLR3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630719.3 | XP_005630776.1 | 11 - 12 | 1382 | 167 | |
| XM_014120581.2 | XP_013976056.1 | 12 - 13 | 1364 | 167 | ||
| XM_022404935.1 | XP_022260643.1 | 10 - 11 | 1277 | 167 | ||
| XM_540277.6 | XP_540277.2 | 12 - 13 | 1475 | 167 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

POLR3C
Genomic Map