Assay Details
| Gene Symbol: HNF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435293.4 | XP_003435341.1 | 4 | 774 | 132 | |
| XM_005624790.3 | XP_005624847.1 | 4 | 852 | 132 | ||
| XM_005624791.3 | XP_005624848.1 | 4 | 774 | 132 | ||
| XM_014116744.2 | XP_013972219.1 | 4 | 981 | 132 | ||
| XM_022423284.1 | XP_022278992.1 | 4 | 1050 | 132 | ||
| XM_022423285.1 | XP_022278993.1 | 4 | 1050 | 132 | ||
| XM_022423286.1 | XP_022278994.1 | 4 | 972 | 132 | ||
| XM_022423287.1 | XP_022278995.1 | 4 | 973 | 132 | ||
| XM_022423288.1 | XP_022278996.1 | 4 | 609 | 132 | ||
| XM_847901.5 | XP_852994.2 | 4 | 852 | 132 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HNF1B
Genomic Map