Assay Details
Gene Symbol: SLC2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159326.1 | NP_001152798.1 | 2 - 3 | 113 | 65 | |
XM_022427664.1 | XP_022283372.1 | 2 - 3 | 178 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
