Assay Details
| Gene Symbol: PTTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417737.1 | XP_022273445.1 | 5 | 510 | 70 | |
| XM_022417738.1 | XP_022273446.1 | 5 | 545 | 70 | ||
| XM_536445.5 | XP_536445.2 | 5 | 548 | 70 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PTTG1
Genomic Map