Assay Details
| Gene Symbol: MAN1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014120656.2 | XP_013976131.1 | 2 - 3 | 981 | 122 | |
| XM_022404884.1 | XP_022260592.1 | 3 - 4 | 981 | 122 | ||
| XM_022404885.1 | XP_022260593.1 | 2 - 3 | 982 | 122 | ||
| XM_540259.6 | XP_540259.2 | 3 - 4 | 948 | 122 | ||
More Information
Set Membership: |
|

MAN1A2
Genomic Map