Assay Details
| Gene Symbol: HOXA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427511.1 | XP_022283219.1 | 2 | 638 | 124 | |
| XM_022427512.1 | XP_022283220.1 | 2 | 816 | 124 | ||
| XM_859585.5 | XP_864678.2 | 2 | 1200 | 124 | ||
More Information
Set Membership: |
|

HOXA7
Genomic Map