Assay Details
| Gene Symbol: ZNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115460.2 | XP_013970935.1 | 3 - 4 | 1036 | 129 | |
| XM_022421465.1 | XP_022277173.1 | 2 - 3 | 775 | 129 | ||
| XM_537285.6 | XP_537285.1 | 2 - 3 | 897 | 129 | ||
| Gene Symbol: ZNF396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421719.1 | XP_022277427.1 | 7 - 8 | 2830 | 129 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZNF24
Genomic Map