Assay Details
| Gene Symbol: TFDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634136.3 | XP_005634193.2 | 3 - 4 | 212 | 95 | |
| XM_022408179.1 | XP_022263887.1 | 3 - 4 | 215 | 95 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TFDP1
Genomic Map