Assay Details
Gene Symbol: NR1H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616270.3 | XP_005616327.1 | 6 - 7 | 1041 | 134 | |
XM_022423354.1 | XP_022279062.1 | 6 - 7 | 1068 | 134 | ||
XM_846223.5 | XP_851316.1 | 6 - 7 | 1036 | 134 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
