Assay Details
| Gene Symbol: PXMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634946.3 | XP_005635003.1 | 1 | 176 | 82 | |
| XM_847386.5 | XP_852479.2 | 1 | 171 | 82 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PXMP4
Genomic Map