Assay Details
| Gene Symbol: LPAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621995.3 | XP_005622052.1 | 3 | 966 | 129 | |
| XM_022420570.1 | XP_022276278.1 | 3 | 984 | 144 | ||
| XM_022420571.1 | XP_022276279.1 | 3 | 929 | 144 | ||
| XM_022420572.1 | XP_022276280.1 | 3 | 1061 | 144 | ||
More Information
Set Membership: |
|

LPAR3
Genomic Map