Assay Details
| Gene Symbol: HARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617292.3 | XP_005617349.1 | 719 | 115 | ||
| XM_005617293.3 | XP_005617350.1 | 468 | 115 | |||
| XM_014107701.2 | XP_013963176.1 | 719 | 115 | |||
| XM_022408565.1 | XP_022264273.1 | 427 | 115 | |||
| XM_535214.6 | XP_535214.3 | 680 | 115 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HARS2
Genomic Map