Assay Details
| Gene Symbol: FADS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631649.3 | XP_005631706.1 | 5 - 6 | 567 | 185 | |
| XM_014120903.2 | XP_013976378.1 | 5 - 6 | 821 | 185 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FADS1
Genomic Map