Assay Details
| Gene Symbol: REM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634909.3 | XP_005634966.1 | 5 | 876 | 156 | |
| XM_542947.4 | XP_542947.1 | 5 | 886 | 156 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: REM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634909.3 | XP_005634966.1 | 5 | 876 | 156 | |
| XM_542947.4 | XP_542947.1 | 5 | 886 | 156 | ||
Set Membership: |
|