Assay Details
| Gene Symbol: RNF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629010.3 | XP_005629067.1 | 3 | 338 | 82 | |
| XM_014119683.2 | XP_013975158.1 | 3 | 347 | 82 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RNF11
Genomic Map