Assay Details
| Gene Symbol: NSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417192.1 | XP_022272900.1 | 2 - 3 | 364 | 127 | |
| XM_536243.6 | XP_536243.4 | 2 - 3 | 364 | 127 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NSG1
Genomic Map