Assay Details
| Gene Symbol: BBC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616403.3 | XP_005616460.1 | 3 - 4 | 2215 | 105 | |
| XM_022424594.1 | XP_022280302.1 | 4 - 5 | 625 | 105 | ||
| XM_849883.5 | XP_854976.1 | 3 - 4 | 736 | 105 | ||
More Information
Set Membership: |
|

BBC3
Genomic Map