Assay Details
| Gene Symbol: SLC39A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639152.4 | XP_003639200.1 | 3 - 4 | 463 | 115 | |
| XM_005622808.3 | XP_005622865.1 | 3 - 4 | 443 | 115 | ||
| XM_022421392.1 | XP_022277100.1 | 3 - 4 | 503 | 115 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC39A1
Genomic Map