Assay Details
| Gene Symbol: PPM1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014108210.2 | XP_013963685.1 | 5 - 6 | 836 | 170 | |
| XM_022410172.1 | XP_022265880.1 | 5 - 6 | 808 | 170 | ||
| XM_543574.6 | XP_543574.3 | 5 - 6 | 1011 | 170 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPM1F
Genomic Map