Assay Details
| Gene Symbol: ST18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109260.2 | XP_013964735.1 | 12 - 13 | 2185 | 155 | |
| XM_014109261.2 | XP_013964736.1 | 12 - 13 | 2182 | 155 | ||
| XM_022412469.1 | XP_022268177.1 | 12 - 13 | 2199 | 155 | ||
| XM_022412470.1 | XP_022268178.1 | 12 - 13 | 2202 | 155 | ||
| XM_022412471.1 | XP_022268179.1 | 9 - 10 | 2193 | 155 | ||
More Information
Set Membership: |
|

ST18
Genomic Map