Assay Details
| Gene Symbol: SPCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003324.2 | NP_001003324.1 | 4 | 493 | 167 | |
| GenBank mRNA | U12687.1 | - | 4 | 494 | 167 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPCS2
Genomic Map