Assay Details
| Gene Symbol: TOMM70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414015.1 | XP_022269723.1 | 382 | 109 | ||
| XM_535719.5 | XP_535719.3 | 645 | 109 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TOMM70
Genomic Map