Assay Details
| Gene Symbol: SWAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122796.2 | XP_013978271.1 | 7 - 8 | 1052 | 121 | |
| XM_542503.6 | XP_542503.3 | 8 - 9 | 1283 | 121 | ||
More Information
Set Membership: |
|

SWAP70
Genomic Map