Assay Details
| Gene Symbol: SARM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423336.1 | XP_022279044.1 | 3 - 4 | 1028 | 110 | |
| XM_548285.6 | XP_548285.2 | 3 - 4 | 1334 | 110 | ||
More Information
Set Membership: |
|

SARM1
Genomic Map