Assay Details
| Gene Symbol: GLCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638539.3 | XP_005638596.1 | 4 | 1747 | 128 | |
| XM_022412590.1 | XP_022268298.1 | 5 | 1823 | 128 | ||
| XM_022412591.1 | XP_022268299.1 | 3 | 1151 | 128 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GLCE
Genomic Map