Assay Details
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630308.3 | XP_005630365.1 | 4 - 5 | 539 | 122 | |
XM_022404611.1 | XP_022260319.1 | 3 - 4 | 491 | 122 | ||
XM_532921.4 | XP_532921.1 | 2 - 3 | 289 | 122 | ||
XM_857398.5 | XP_862491.1 | 2 - 3 | 1424 | 122 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
