Assay Details
| Gene Symbol: BOLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639618.4 | XP_003639666.1 | 2 | 478 | 137 | |
| XM_022404949.1 | XP_022260657.1 | 2 | 530 | 137 | ||
| Gene Symbol: H3C15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404950.1 | XP_022260658.1 | 4 | 1173 | 137 | |
| XM_022404951.1 | XP_022260659.1 | 5 | 1325 | 137 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BOLA1
Genomic Map