Assay Details
| Gene Symbol: IER3IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421425.1 | XP_022277133.1 | 4 | 344 | 244 | |
| XM_850110.5 | XP_855203.1 | 2 | 229 | 149 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IER3IP1
Genomic Map