Assay Details
| Gene Symbol: CPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022412248.1 | XP_022267956.1 | 2 | 463 | 108 | |
| XM_022412249.1 | XP_022267957.1 | 2 | 465 | 108 | ||
| XM_022412250.1 | XP_022267958.1 | 2 | 465 | 108 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CPQ
Genomic Map