Assay Details
| Gene Symbol: SDF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620370.3 | XP_005620427.1 | 577 | 140 | ||
| XM_005620372.3 | XP_005620429.1 | 369 | 140 | |||
| XM_022418890.1 | XP_022274598.1 | 574 | 140 | |||
| XM_536712.6 | XP_536712.3 | 381 | 140 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SDF4
Genomic Map