Assay Details
| Gene Symbol: HECA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417566.1 | XP_022273274.1 | 1172 | 180 | ||
| XM_022417569.1 | XP_022273277.1 | 1302 | 180 | |||
| XM_022417573.1 | XP_022273281.1 | 1302 | 180 | |||
| XM_022417577.1 | XP_022273285.1 | 1197 | 180 | |||
| XM_022417578.1 | XP_022273286.1 | 1191 | 180 | |||
| XM_533428.6 | XP_533428.2 | 1208 | 180 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HECA
Genomic Map