Assay Details
| Gene Symbol: CCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116544.2 | XP_013972019.1 | 1095 | 121 | ||
| XM_844228.5 | XP_849321.1 | 887 | 121 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116544.2 | XP_013972019.1 | 1095 | 121 | ||
| XM_844228.5 | XP_849321.1 | 887 | 121 | |||
Set Membership: |
|