Assay Details
| Gene Symbol: MAP3K20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414748.1 | XP_022270456.1 | 1 - 2 | 197 | 76 | |
| XM_022414749.1 | XP_022270457.1 | 1 - 2 | 305 | 76 | ||
| XM_022414750.1 | XP_022270458.1 | 1 - 2 | 197 | 76 | ||
| XM_535966.6 | XP_535966.2 | 2 - 3 | 448 | 76 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAP3K20
Genomic Map